Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSDMC
Entrez Gene ID
101103902
Gene Name
gasdermin C
Gene Aliases
-
Chromosome Location
Chr.9: 25589254 - 25607175 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972904.1 XP_027828705.1 13 - 14 1471 85

View Full Product Details