Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIER1
Entrez Gene ID
101113806
Gene Name
MIER1 transcriptional regulator
Gene Aliases
-
Chromosome Location
Chr.1: 44789006 - 44855276 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015092042.2 XP_014947528.2 11 - 12 1020 87

View Full Product Details