Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCGB1C1
Entrez Gene ID
101105959
Gene Name
secretoglobin family 1C member 1
Gene Aliases
-
Chromosome Location
Chr.21: 52822100 - 52823988 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019746.4 XP_004019795.2 1 - 2 459 83

View Full Product Details