Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PARP10
Entrez Gene ID
101111201
Gene Name
poly(ADP-ribose) polymerase family member 10
Gene Aliases
-
Chromosome Location
Chr.9: 15037863 - 15044846 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973336.1 XP_027829137.1 7 - 8 2495 76

View Full Product Details