Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM83B
Entrez Gene ID
101108170
Gene Name
family with sequence similarity 83 member B
Gene Aliases
-
Chromosome Location
Chr.20: 7028382 - 7112173 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958449.1 XP_027814250.1 1 - 2 508 105

View Full Product Details