Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP300
Entrez Gene ID
101121882
Gene Name
cilia and flagella associated protein 300
Gene Aliases
C15H11orf70
Chromosome Location
Chr.15: 7036010 - 7064710 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979145.1 XP_027834946.1 4 - 5 465 135

View Full Product Details