Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP1LC3C
Entrez Gene ID
101120002
Gene Name
microtubule associated protein 1 light chain 3 gamma
Gene Aliases
-
Chromosome Location
Chr.12: 38133572 - 38139286 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012187218.3 XP_012042608.1 1 - 2 131 66

View Full Product Details