Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CATSPERG
Entrez Gene ID
101111312
Gene Name
cation channel sperm associated auxiliary subunit gamma
Gene Aliases
-
Chromosome Location
Chr.14: 51027833 - 51060775 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978237.1 XP_027834038.1 19 - 20 4323 107

View Full Product Details