Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFA2
Entrez Gene ID
101119470
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Gene Aliases
-
Chromosome Location
Chr.5: 54137360 - 54139494 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008868.3 XP_004008917.1 2 - 3 253 114

View Full Product Details