Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC15
Entrez Gene ID
101109059
Gene Name
sialic acid binding Ig like lectin 15
Gene Aliases
-
Chromosome Location
Chr.23: 51013109 - 51030168 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960801.1 XP_027816602.1 1 - 2 178 74

View Full Product Details