Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOR1B
Entrez Gene ID
101103637
Gene Name
torsin family 1 member B
Gene Aliases
-
Chromosome Location
Chr.3: 6858246 - 6865298 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966306.1 XP_027822107.1 3 - 4 729 75

View Full Product Details