Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTL3
Entrez Gene ID
101106496
Gene Name
retrotransposon Gag like 3
Gene Aliases
ZCCHC5
Chromosome Location
Chr.X: 72475726 - 72477444 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104713.1 XP_014960199.1 2 - 2 1423 97

View Full Product Details