Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HESX1
Entrez Gene ID
100008708
Gene Name
HESX homeobox 1
Gene Aliases
ANF
Chromosome Location
Chr.9: 24073422 - 24075869 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002713403.3 XP_002713449.1 3 - 4 780 112
GenBank mRNA AF102132.1 - 3 - 4 552 112

View Full Product Details