Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNAS
Entrez Gene ID
100008904
Gene Name
GNAS complex locus
Gene Aliases
G-SALPHA60A
Chromosome Location
Chr.Unknown: 265709 - 315518 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002721804.2 XP_002721850.1 - 2195 70
XM_008248810.2 XP_008247032.1 - 2150 70
XM_008248812.2 XP_008247034.2 - 311 70
XM_008248816.2 XP_008247038.1 - 266 70
XM_017337815.1 XP_017193304.1 - 2198 70
XM_017337816.1 XP_017193305.1 - 2153 70
GenBank mRNA AY248719.1 - - 271 70

View Full Product Details