Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPD52
Entrez Gene ID
100009212
Gene Name
tumor protein D52
Gene Aliases
-
Chromosome Location
Chr.3: 95776594 - 95797952 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082360.1 NP_001075829.1 2 - 3 138 76
GenBank mRNA U35428.1 - 2 - 3 138 76

View Full Product Details