Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG1
Entrez Gene ID
100009263
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 1
Gene Aliases
-
Chromosome Location
Chr.19: 50421353 - 50431417 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082393.1 NP_001075862.1 3 - 4 577 61
XM_008271101.2 XP_008269323.1 4 - 5 743 61
GenBank mRNA M32231.1 - 3 - 4 484 61
X56031.1 - - 577 61

View Full Product Details