Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RYR2
Entrez Gene ID
100009536
Gene Name
ryanodine receptor 2
Gene Aliases
RYR-2
Chromosome Location
Chr.16: 27050052 - 27608942 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082757.1 NP_001076226.1 57 - 58 8901 122
GenBank mRNA M59743.1 - 57 - 58 8891 122
U50465.1 - 57 - 58 8901 122

View Full Product Details