The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122939.1 | NP_001116411.1 | 2 - 3 | 325 | 91 |
XM_017341620.1 | XP_017197109.1 | 2 - 3 | 548 | 91 | |
GenBank mRNA | AM748787.1 | - | 2 - 3 | 325 | 91 |
FN547132.1 | - | 2 - 3 | 294 | 91 | |
FN547133.1 | - | 2 - 3 | 220 | 91 | |
FN547134.1 | - | 1 - 2 | 315 | 91 |