Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEL
Entrez Gene ID
100009339
Gene Name
carboxyl ester lipase
Gene Aliases
-
Chromosome Location
Chr.Unknown: 461 - 4614 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101711.2 NP_001095181.3 - 1373 101
XM_017340098.1 XP_017195587.1 - 1465 101
GenBank mRNA X65944.1 - - 1349 101

View Full Product Details