Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA1
Entrez Gene ID
100009083
Gene Name
carbonic anhydrase 1
Gene Aliases
CAR1
Chromosome Location
Chr.3: 100976946 - 101025544 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017341428.1 XP_017196917.1 - 592 101
GenBank mRNA M10412.1 - - 154 101

View Full Product Details