Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP7A1
Entrez Gene ID
100328551
Gene Name
cytochrome P450, family 7, subfamily A, polypeptide 1
Gene Aliases
CYP7
Chromosome Location
Chr.3: 73947427 - 73958448 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170929.1 NP_001164400.1 5 - 6 1286 96
GenBank mRNA L10754.1 - 5 - 6 1286 96

View Full Product Details