The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171124.1 | NP_001164595.1 | 1 - 1 | 946 | 101 |
XM_008260477.2 | XP_008258699.1 | 2 - 2 | 1327 | 101 |