Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP1A
Entrez Gene ID
100328681
Gene Name
microtubule associated protein 1A
Gene Aliases
-
Chromosome Location
Chr.17: 28723892 - 28744156 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171311.1 NP_001164782.1 1 - 2 4200 70

View Full Product Details