Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPAG4
Entrez Gene ID
100328707
Gene Name
sperm associated antigen 4
Gene Aliases
-
Chromosome Location
Chr.4: 4216813 - 4220543 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171332.1 NP_001164803.1 1 - 2 307 93

View Full Product Details