Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFC
Entrez Gene ID
100328738
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
Chromosome Location
Chr.4: 88630325 - 88697818 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171359.1 NP_001164830.1 3 - 4 371 64
XM_008257060.2 XP_008255282.1 4 - 5 479 64
XM_008257061.2 XP_008255283.1 5 - 6 1825 64
XM_008257062.2 XP_008255284.1 3 - 4 351 64

View Full Product Details