Gene Transcripts

Gene Symbol
Slfn3
Entrez Gene ID
114247
Gene Name
schlafen 3
Gene Aliases
Cdk107, Slfn4
Chromosome Location
Chr. 10: 70411688 - 70435162 on Build Rnor_6.0
UniGene ID
Rn.80831
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053687.1 NP_446139.1 6 3062
XM_008767933.1 XP_008766155.1 8 3524
GenBank mRNA AF168795.1 - 6 3062

View Full Product Details