Gene Transcripts

Gene Symbol
Atp8a2
Entrez Gene ID
691889
Gene Name
ATPase, aminophospholipid transporter, class I, type 8A, member 2
Gene Aliases
Chromosome Location
Chr. 15: 39964486 - 40468325 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751492.3 XP_003751540.1 8 2312
XM_006252160.2 XP_006252222.1 8 979
XM_006252164.2 XP_006252226.1 8 1850
XM_006252165.2 XP_006252227.1 8 584
XM_008770789.1 XP_008769011.1 8 1723
XM_008770790.1 XP_008769012.1 8 1040
XM_008770791.1 XP_008769013.1 9 708

View Full Product Details