Gene Transcripts

Gene Symbol
Gpr33
Entrez Gene ID
299007
Gene Name
G protein-coupled receptor 33
Gene Aliases
Chromosome Location
Chr. 6: 72804843 - 72805848 on Build Rnor_6.0
UniGene ID
Rn.152676
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031823.1 NP_001026993.1 2 358

View Full Product Details