Gene Transcripts

Gene Symbol
Zfp39
Entrez Gene ID
303173
Gene Name
zinc finger protein 39
Gene Aliases
Chromosome Location
Chr. 10: 45220352 - 45243653 on Build Rnor_6.0
UniGene ID
Rn.163096
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107004.2 NP_001100474.2 5 1078

View Full Product Details