Gene Transcripts

Gene Symbol
Ccdc19
Entrez Gene ID
304984
Gene Name
coiled-coil domain containing 19
Gene Aliases
Chromosome Location
Chr. 13: 90909480 - 90932958 on Build Rnor_6.0
UniGene ID
Rn.137285
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024882.1 NP_001020053.1 9 1162
XM_006250296.2 XP_006250358.1 9 1257
GenBank mRNA BC098058.1 - 9 1162

View Full Product Details