Gene Transcripts

Gene Symbol
Mettl17
Entrez Gene ID
305845
Gene Name
methyltransferase like 17
Gene Aliases
Mett11d1, RGD1559895
Chromosome Location
Chr. 15: 28287033 - 28293869 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_223974.8 XP_223974.4 3 526

View Full Product Details