Gene Transcripts

Gene Symbol
Tmem8b
Entrez Gene ID
313490
Gene Name
transmembrane protein 8B
Gene Aliases
RGD1310012
Chromosome Location
Chr. 5: 59165712 - 59193772 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238139.2 XP_006238201.1 11 2338
XM_008763686.1 XP_008761908.1 9 3476
XM_008763688.1 XP_008761910.1 12 1864

View Full Product Details