Gene Transcripts

Gene Symbol
Sbno2
Entrez Gene ID
314619
Gene Name
strawberry notch homolog 2 (Drosophila)
Gene Aliases
RGD1307194, Stno
Chromosome Location
Chr. 7: 12468909 - 12515700 on Build Rnor_6.0
UniGene ID
Rn.67287
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108068.1 NP_001101538.1 19 2330
XM_006240908.2 XP_006240970.1 18 3612
XM_006240909.1 XP_006240971.1 16 3073
XM_008765094.1 XP_008763316.1 18 2195
XM_008765095.1 XP_008763317.1 15 3758

View Full Product Details