Gene Transcripts

Gene Symbol
Ibtk
Entrez Gene ID
315858
Gene Name
inhibitor of Bruton agammaglobulinemia tyrosine kinase
Gene Aliases
Chromosome Location
Chr. 8: 93323454 - 93397047 on Build Rnor_6.0
UniGene ID
Rn.35504
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766469.1 XP_008764691.1 18, 19 4394

View Full Product Details