Gene Transcripts

Gene Symbol
Ing5
Entrez Gene ID
363292
Gene Name
inhibitor of growth family, member 5
Gene Aliases
Chromosome Location
Chr. 9: 100932932 - 100950233 on Build Rnor_6.0
UniGene ID
Rn.214030
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108810.1 NP_001102280.1 3 268
GenBank mRNA BC167048.1 - 3 139

View Full Product Details