Gene Transcripts

Gene Symbol
Clec2l
Entrez Gene ID
296985
Gene Name
C-type lectin domain family 2, member L
Gene Aliases
RGD1565373
Chromosome Location
Chr. 4: 66405166 - 66421654 on Build Rnor_6.0
UniGene ID
Rn.162737
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044233.2 NP_001037698.2 3, 4 415
GenBank mRNA BC161812.1 - 545
DQ630549.1 - 3, 4 275

View Full Product Details