Gene Transcripts

Gene Symbol
Nova1
Entrez Gene ID
298992
Gene Name
neuro-oncological ventral antigen 1
Gene Aliases
Nova-1
Chromosome Location
Chr. 6: 66960126 - 67085844 on Build Rnor_6.0
UniGene ID
Rn.206187
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100541.1 NP_001094011.1 4 900
XM_006240069.2 XP_006240131.1 5 1426
XM_006240070.2 XP_006240132.1 5 1310
XM_006240072.2 XP_006240134.1 4 2133
GenBank mRNA AY262017.1 - 4 900

View Full Product Details