Gene Transcripts

Gene Symbol
Cpg1
Entrez Gene ID
299247
Gene Name
candidate plasticity gene 1
Gene Aliases
Chromosome Location
Chr. 6: 123034304 - 123046297 on Build Rnor_6.0
UniGene ID
Rn.10347
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178104.2 NP_835205.1 2 987
GenBank mRNA X73292.1 - 2 987

View Full Product Details