Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_212519.2 | NP_997684.1 | 4 | 822 | |
XM_008768643.1 | XP_008766865.1 | 4 | 1116 | ||
XM_008768644.1 | XP_008766866.1 | 4 | 1122 | ||
XM_008768645.1 | XP_008766867.1 | 4 | 1120 | ||
GenBank mRNA | AB162856.1 | - | 4 | 685 | |
BC098640.1 | - | 4 | 822 | 7099109 | |
FQ228582.1 | - | 867 |