Gene Transcripts

Gene Symbol
Ssc4d
Entrez Gene ID
304401
Gene Name
scavenger receptor cysteine rich family, 4 domains
Gene Aliases
Srcrb4d
Chromosome Location
Chr. 12: 23747646 - 23768880 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249209.2 XP_006249271.1 2 539
XM_006249211.2 XP_006249273.1 2 539
XM_222076.8 XP_222076.5 2 504

View Full Product Details