Gene Transcripts

Gene Symbol
Imp3
Entrez Gene ID
315697
Gene Name
IMP3, U3 small nucleolar ribonucleoprotein
Gene Aliases
RGD1306825
Chromosome Location
Chr. 8: 61607021 - 61607909 on Build Rnor_6.0
UniGene ID
Rn.3787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108152.1 NP_001101622.1 1 507
GenBank mRNA BC166985.1 - 513

View Full Product Details