Gene Transcripts

Gene Symbol
Clec4m
Entrez Gene ID
363856
Gene Name
C-type lectin domain family 4, member M
Gene Aliases
Cd209d, Dc-signr, RGD1561104
Chromosome Location
Chr. 12: 2406879 - 2413324 on Build Rnor_6.0
UniGene ID
Rn.218629
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108849.1 NP_001102319.1 7 732

View Full Product Details