Gene Transcripts

Gene Symbol
Ascl5
Entrez Gene ID
363991
Gene Name
achaete-scute family bHLH transcription factor 5
Gene Aliases
RGD1561854
Chromosome Location
Chr. 13: 52887649 - 52888215 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769577.1 XP_008767799.1 1 156

View Full Product Details