Gene Transcripts

Gene Symbol
Vom1r83
Entrez Gene ID
494301
Gene Name
vomeronasal 1 receptor 83
Gene Aliases
V1rc18
Chromosome Location
Chr. 4: 88184956 - 88185894 on Build Rnor_6.0
UniGene ID
Rn.129657
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008956.1 NP_001008956.1 1 341
GenBank mRNA AY510314.1 - 1 341

View Full Product Details