Gene Transcripts

Gene Symbol
Fgfbp3
Entrez Gene ID
499349
Gene Name
fibroblast growth factor binding protein 3
Gene Aliases
RGD1560326
Chromosome Location
Chr. 1: 255555050 - 255557055 on Build Rnor_6.0
UniGene ID
Rn.166095
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109165.1 NP_001102635.1 2 575

View Full Product Details