Gene Transcripts

Gene Symbol
Vom2r50
Entrez Gene ID
500312
Gene Name
vomeronasal 2 receptor, 50
Gene Aliases
Chromosome Location
Chr. 4: 156482446 - 156504818 on Build Rnor_6.0
UniGene ID
Rn.134207
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099507.1 NP_001092977.1 3 1294

View Full Product Details