Gene Transcripts

Gene Symbol
Hephl1
Entrez Gene ID
500946
Gene Name
hephaestin-like 1
Gene Aliases
RGD1564835
Chromosome Location
Chr. 8: 13614800 - 13680870 on Build Rnor_6.0
UniGene ID
Rn.228190
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191993.1 NP_001178922.1 1 27

View Full Product Details