Gene Transcripts

Gene Symbol
Vom2r43
Entrez Gene ID
679593
Gene Name
vomeronasal 2 receptor, 43
Gene Aliases
Chromosome Location
Chr. 1: 149506822 - 149529350 on Build Rnor_6.0
UniGene ID
Rn.217927
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099512.1 NP_001092982.1 2 377

View Full Product Details