Gene Transcripts

Gene Symbol
Tead4
Entrez Gene ID
683522
Gene Name
TEA domain family member 4
Gene Aliases
Chromosome Location
Chr. 4: 161540110 - 161619842 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006237424.2 XP_006237486.1 3 2321

View Full Product Details