Gene Transcripts

Gene Symbol
Ttll3
Entrez Gene ID
362415
Gene Name
tubulin tyrosine ligase-like family, member 3
Gene Aliases
Chromosome Location
Chr. 4: 145342154 - 145366083 on Build Rnor_6.0
UniGene ID
Rn.229484
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108640.2 NP_001102110.2 3 216
GenBank mRNA BC168907.1 - 2 74
CV126544.1 - 3 326

View Full Product Details